Thermo Scientific Chemicals

N,N'-Di-n-butylthiourea, 98%, Thermo Scientific Chemicals

Catalog number: A16783.18
50 g, Each
Thermo Scientific Chemicals

N,N'-Di-n-butylthiourea, 98%, Thermo Scientific Chemicals

Catalog number: A16783.18
50 g, Each
Quantity
Catalog number: A16783.18
also known as A16783-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
109-46-6
IUPAC Name
1,3-dibutylthiourea
Molecular Formula
C9H20N2S
InChI Key
KFFQABQEJATQAT-UHFFFAOYSA-N
SMILES
CCCCNC(=S)NCCCC
Appearance (Color)
White to cream or yellow
Form
Crystals or powder or crystalline powder
Assay (HPLC)
≥97.5%
Melting Point (clear melt)
62.0-68.0?C

Description

N,N'-Di-n-butylthiourea is used in vulcanization accelerator and corrosion inhibitors for metal treatments. It is used as leather, plastic and rubber auxiliary agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N′-Di-n-butylthiourea is used in vulcanization accelerator and corrosion inhibitors for metal treatments. It is used as leather, plastic and rubber auxiliary agents.

Solubility
Insoluble in water.

Notes
Store in a cool, dry, conditions in well sealed container. Store away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text