N,N'-Di-n-butylthiourea is used in vulcanization accelerator and corrosion inhibitors for metal treatments. It is used as leather, plastic and rubber auxiliary agents.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
N,N′-Di-n-butylthiourea is used in vulcanization accelerator and corrosion inhibitors for metal treatments. It is used as leather, plastic and rubber auxiliary agents.
Solubility
Insoluble in water.
Notes
Store in a cool, dry, conditions in well sealed container. Store away from oxidizing agents.
RUO – Research Use Only
Jussi Liippo; Leena Ackermann; Taina Hasan; Arja Laukkanen; Tapio Rantanen; Kaija Lammintausta. Sensitization to thiourea derivatives among Finnish patients with suspected contact dermatitis. Contact Dermatitis.2010, 63 (1), 37-41.