Synthesis of quinoline-based potential anticancer agents. Substrate for palladium-catalyzed coupling reactions
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Synthesis of quinoline-based potential anticancer agents. Substrate for palladium-catalyzed coupling reactions
Solubility
Easily soluble in water.
Notes
Light sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only
General References:
- Michael Lewis,; Charles L. Barnes,; Rainer Glaser.The crystal structure of 4-iodoacetophenone azine. Journal of Chemical Crystallography. 1999, 29 (9), 1043-1048.
- Michael Lewis,; Charles L. Barnes,; Rainer Glaser. Near-perfect dipole parallel-alignment in the highly anisotropic crystal structure of 4-iodoacetophenone-(4-methoxyphenylethylidene) hydrazone. Journal of Chemical Crystallography. 2000, 30(7), 489-496 .