1-Nitro-3,5-bis(trifluoromethyl)benzene, 96%, Thermo Scientific Chemicals
1-Nitro-3,5-bis(trifluoromethyl)benzene, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1-Nitro-3,5-bis(trifluoromethyl)benzene, 96%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number A16797.06
also known as A16797-06
Price (USD)/ Each
36.10
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
36.10
Add to cart
1-Nitro-3,5-bis(trifluoromethyl)benzene, 96%, Thermo Scientific Chemicals
Catalog numberA16797.06
Price (USD)/ Each
36.10
-
Add to cart
Chemical Identifiers
CAS328-75-6
IUPAC Name1-nitro-3,5-bis(trifluoromethyl)benzene
Molecular FormulaC8H3F6NO2
InChI KeyGMUWJDVVXLBMEZ-UHFFFAOYSA-N
SMILES[O-][N+](=O)C1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear yellow to orange
Refractive Index1.4240-1.4300 @ 20°C
FormLiquid
Assay (GC)≥95.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only