S-Acetylthiocholine iodide, 98%, Thermo Scientific Chemicals
S-Acetylthiocholine iodide, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

S-Acetylthiocholine iodide, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
25 g
Catalog number A16802.06
also known as A16802-06
Price (USD)/ Each
78.65
Online exclusive
87.20 
Save 8.55 (10%)
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
78.65
Online exclusive
87.20 
Save 8.55 (10%)
Add to cart
S-Acetylthiocholine iodide, 98%, Thermo Scientific Chemicals
Catalog numberA16802.06
Price (USD)/ Each
78.65
Online exclusive
87.20 
Save 8.55 (10%)
-
Add to cart
Chemical Identifiers
CAS1866-15-5
IUPAC Name[2-(acetylsulfanyl)ethyl]trimethylazanium iodide
Molecular FormulaC7H16INOS
InChI KeyNTBLZMAMTZXLBP-UHFFFAOYSA-M
SMILES[I-].CC(=O)SCC[N+](C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormCrystals or powder or crystalline powder
Assay (Titration ex Iodide)≥97.5 to ≤102.5%
Identification (FTIR)Conforms
Water Content (Karl Fischer Titration)≤1.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only