Thermo Scientific Chemicals

Bis(4-aminophenyl) ether, 98%, Thermo Scientific Chemicals

Catalog number: A16815.18
50 g, Each
Thermo Scientific Chemicals

Bis(4-aminophenyl) ether, 98%, Thermo Scientific Chemicals

Catalog number: A16815.18
50 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: A16815.18
also known as A16815-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
101-80-4
IUPAC Name
4-(4-aminophenoxy)aniline
Molecular Formula
C12H12N2O
InChI Key
HLBLWEWZXPIGSM-UHFFFAOYSA-N
SMILES
NC1=CC=C(OC2=CC=C(N)C=C2)C=C1
Form
Crystals or powder or crystalline powder
Melting Point
187-195°C
Appearance (Color)
White to cream to pale brown
Assay (Non-aqueous acid-base Titration)
≥97.5 to ≤102.5%
Identification (FTIR)
Conforms

Description

Bis(4-aminophenyl) ether is a monomer used as a cross-linking agent. It is also used in the preparation of poly(imideurea)s, polyimide silica composite membrane, which finds application in gas separation techniques. It is utilized in products such as wire enamels, coatings, adhesives, insulating varnishes, coated fabrics, flame-retardant fibers, oil sealants and retainers. Further, it is used in the preparation of polymer films for electrolyte membranes. In addition to this, it is used as an insulation for cables and printed circuits, and laminates and composite for aerospace vehicles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bis(4-aminophenyl) ether is a monomer used as a cross-linking agent. It is also used in the preparation of poly(imideurea)s, polyimide silica composite membrane, which finds application in gas separation techniques. It is utilized in products such as wire enamels, coatings, adhesives, insulating varnishes, coated fabrics, flame-retardant fibers, oil sealants and retainers. Further, it is used in the preparation of polymer films for electrolyte membranes. In addition to this, it is used as an insulation for cables and printed circuits, and laminates and composite for aerospace vehicles.

Solubility
Soluble in acetone. Insoluble in water, benzene, carbon tetrachloride, ethanol and hot methanol.

Notes
Hygroscopic. Incompatible with strong oxidizing agents.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text