Bis(4-aminophenyl) ether, 98%, Thermo Scientific Chemicals
Bis(4-aminophenyl) ether, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Bis(4-aminophenyl) ether, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
50 g
500 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number A16815.22
also known as A16815-22
Price (USD)/ Each
53.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
63.50 
Save 9.85 (16%)
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
53.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
63.50 
Save 9.85 (16%)
Add to cart
Bis(4-aminophenyl) ether, 98%, Thermo Scientific Chemicals
Catalog numberA16815.22
Price (USD)/ Each
53.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
63.50 
Save 9.85 (16%)
-
Add to cart
Chemical Identifiers
CAS101-80-4
IUPAC Name4-(4-aminophenoxy)aniline
Molecular FormulaC12H12N2O
InChI KeyHLBLWEWZXPIGSM-UHFFFAOYSA-N
SMILESNC1=CC=C(OC2=CC=C(N)C=C2)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Melting Point187-195°C
Appearance (Color)White to cream to pale brown
Assay (Non-aqueous acid-base Titration)≥97.5 to ≤102.5%
Identification (FTIR)Conforms
View more
Bis(4-aminophenyl) ether is a monomer used as a cross-linking agent. It is also used in the preparation of poly(imideurea)s, polyimide silica composite membrane, which finds application in gas separation techniques. It is utilized in products such as wire enamels, coatings, adhesives, insulating varnishes, coated fabrics, flame-retardant fibers, oil sealants and retainers. Further, it is used in the preparation of polymer films for electrolyte membranes. In addition to this, it is used as an insulation for cables and printed circuits, and laminates and composite for aerospace vehicles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bis(4-aminophenyl) ether is a monomer used as a cross-linking agent. It is also used in the preparation of poly(imideurea)s, polyimide silica composite membrane, which finds application in gas separation techniques. It is utilized in products such as wire enamels, coatings, adhesives, insulating varnishes, coated fabrics, flame-retardant fibers, oil sealants and retainers. Further, it is used in the preparation of polymer films for electrolyte membranes. In addition to this, it is used as an insulation for cables and printed circuits, and laminates and composite for aerospace vehicles.

Solubility
Soluble in acetone. Insoluble in water, benzene, carbon tetrachloride, ethanol and hot methanol.

Notes
Hygroscopic. Incompatible with strong oxidizing agents.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

General References:

  1. Pethrick, R. A.; Santamaria-Mendia, F. Gas permeation and positron annihilation lifetime spectroscopy of poly(ether imides) containing main chain ethylene oxide segments. J. Polym. Sci., Part B: Polym. Phys. 2015, 53 (23), 1654-1662.
  2. Kaya, İ.; Kartal, E.; Şenol, D. Synthesis and characterization of polyphenol derived from Schiff bases containing methyl and carboxyl groups in the structure. Des. Monomers Polym. 2015, 18 (6), 524-535.