Thermo Scientific Chemicals

(1-Bromoethyl)benzene, 97%, Thermo Scientific Chemicals

Catalog number: A16839.36
500 g, Each
Thermo Scientific Chemicals

(1-Bromoethyl)benzene, 97%, Thermo Scientific Chemicals

Catalog number: A16839.36
500 g, Each
Quantity
Catalog number: A16839.36
also known as A16839-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
585-71-7
IUPAC Name
(1-bromoethyl)benzene
Molecular Formula
C8H9Br
InChI Key
CRRUGYDDEMGVDY-UHFFFAOYNA-N
SMILES
CC(Br)C1=CC=CC=C1
Assay (GC)
≥96.0%
Form
Liquid
Identification (FTIR)
Conforms
Appearance (Color)
Clear colorless to pale yellow to yellow to brown to gray
Refractive Index
1.5585-1.5635 @ 20?C

Description

(1-Bromoethyl)benzene has been employed in controlled radical polymerization of styrene, in asymmetric esterification of benzoic acid in the presence of a chiral cyclic guanidine and as initiator in the synthesis of bromine terminated polyp-methoxystyrene and polystyrene via atom transfer radical polymerization.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(1-Bromoethyl)benzene has been employed in controlled radical polymerization of styrene, in asymmetric esterification of benzoic acid in the presence of a chiral cyclic guanidine and as initiator in the synthesis of bromine terminated polyp-methoxystyrene and polystyrene via atom transfer radical polymerization.

Solubility
Soluble in alcohol, ether, and benzene.

Notes
Moisture Sensitive. Do not store together with strongly basic and oxidizing materials.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text