Thermo Scientific Chemicals

3,3-Dimethylacrylic acid, 98%, Thermo Scientific Chemicals

Catalog number: A16865.14
25 g, Each
Thermo Scientific Chemicals

3,3-Dimethylacrylic acid, 98%, Thermo Scientific Chemicals

Catalog number: A16865.14
25 g, Each
Quantity
Catalog number: A16865.14
also known as A16865-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
541-47-9
IUPAC Name
3-methylbut-2-enoic acid
Molecular Formula
C5H8O2
InChI Key
YYPNJNDODFVZLE-UHFFFAOYSA-N
SMILES
CC(C)=CC(O)=O
Assay (Silylated GC)
≥97.5%
Melting Point (clear melt)
64.5-70.5?C
Appearance (Color)
White to pale yellow or cream
Assay (Aqueous acid-base Titration)
≥97.5 to ≤102.5%
Identification (FTIR)
Conforms

Description

Chemical Intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Chemical Intermediates.

Solubility
Soluble in water, and methanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text