Chemical Intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Chemical Intermediates.
Solubility
Soluble in water, and methanol.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from bases.
RUO – Research Use Only
David G. Anderson,; David W. Norgard,; John W. Porter.The incorporation of mevalonic acid-2-C14 and dimethylacrylic acid-3-C14 into carotenes. Archives of Biochemistry and Biophysics. year of publication , 88 (1), 68-77.
PJ Collin,; S Sternhell .N.M.R. spectra of crotonic, mesaconic, citraconic, and β,β-dimethylacrylic acids, their methyl esters and potassium salts. Australian Journal of Chemistry . 1966, 19 (2),317 - 320 .