Thermo Scientific Chemicals

Naphthalene-2,6-dicarboxylic acid, 98+%, Thermo Scientific Chemicals

Catalog number: A16868.22
100 g, Each
Thermo Scientific Chemicals

Naphthalene-2,6-dicarboxylic acid, 98+%, Thermo Scientific Chemicals

Catalog number: A16868.22
100 g, Each
Quantity
Catalog number: A16868.22
also known as A16868-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1141-38-4
IUPAC Name
naphthalene-2,6-dicarboxylic acid
Molecular Formula
C12H8O4
InChI Key
RXOHFPCZGPKIRD-UHFFFAOYSA-N
SMILES
OC(=O)C1=CC2=CC=C(C=C2C=C1)C(O)=O
Appearance (Color)
White to cream
Assay (Silylated GC)
≥98.0%
Assay (Aqueous acid-base Titration)
≥98.0 to ≤102.0%
Form
Powder
Assay (HPLC)
≥98.0%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text