3-Chloro-1-propanol, 98%, stab., Thermo Scientific Chemicals
3-Chloro-1-propanol, 98%, stab., Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Chloro-1-propanol, 98%, stab., Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
Catalog number A16871.36
also known as A16871-36
Price (USD)/ Each
225.65
Online exclusive
250.00 
Save 24.35 (10%)
-
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)/ Each
225.65
Online exclusive
250.00 
Save 24.35 (10%)
Add to cart
3-Chloro-1-propanol, 98%, stab., Thermo Scientific Chemicals
Catalog numberA16871.36
Price (USD)/ Each
225.65
Online exclusive
250.00 
Save 24.35 (10%)
-
Add to cart
Chemical Identifiers
CAS627-30-5
IUPAC Name3-chloropropan-1-ol
Molecular FormulaC3H7ClO
InChI KeyLAMUXTNQCICZQX-UHFFFAOYSA-N
SMILESOCCCCl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
Refractive Index1.4440-1.4500 @ 20?C
FormLiquid
Identification (FTIR)Conforms
Assay (GC)≥97.5%
View more
It is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a pharmaceutical intermediate.

Solubility
Soluble in water at 20°C 300g/L.

Notes
Store away from oxidizing agents and bases. Keep the container tightly sealed and store in cool, dry condition in well ventilated place.
RUO – Research Use Only

General References:

  1. Michael J. Fuller.; E.Bright Wilson.; Walther Caminatia. Rotational isomerism in 3-chloro-1-propanol from the microwave spectrum.J. Mol. Spectrosc.1982, 96(1), 131-145.
  2. N. Baggett.' Md. Mosihuzzaman.; J.M. Webber. Benzylidenation in basic medium: the reation of methyl 2,3-di-O-methyl-6-O-toluene-p-sulphonyl-α-d-glucopyranoside and 3-chloro-1-propanol with benzaldehyde.Carbohydr. Res.1969, 11(2), 263-266.