Thermo Scientific Chemicals

Ferrocenecarboxylic acid, 97+%, Thermo Scientific Chemicals

Catalog number: A16874.06
5 g, Each
Thermo Scientific Chemicals

Ferrocenecarboxylic acid, 97+%, Thermo Scientific Chemicals

Catalog number: A16874.06
5 g, Each
Quantity
Catalog number: A16874.06
Price (USD)
Have Questions?

Chemical Identifiers

CAS
1271-42-7
IUPAC Name
Ferrocenecarboxylic acid
Molecular Formula
C11H10FeO2
SMILES
[Fe].c1cccc1.OC(=O)c1cccc1
Molecular Weight (g/mol)
230.04
Form
Powder
Appearance (Color)
Orange to brown
Assay (Aqueous acid-base Titration)
≥97.0 to ≤103.0%

Description

Ferrocenecarboxylic acid is used to esterify complex mixtures of phenols and alcohols for analysis by GCMS. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ferrocenecarboxylic acid is used to esterify complex mixtures of phenols and alcohols for analysis by GCMS. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.

Solubility
Insoluble in water.

Notes
Store in a cool, dry, conditions in well sealed container. Store away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text