Thermo Scientific Chemicals

4-Iodo-m-xylene, 98%, Thermo Scientific Chemicals

Catalog number: A16878.09
10 g, Each
Thermo Scientific Chemicals

4-Iodo-m-xylene, 98%, Thermo Scientific Chemicals

Catalog number: A16878.09
10 g, Each
Quantity
Catalog number: A16878.09
also known as A16878-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
4214-28-2
IUPAC Name
1-iodo-2,4-dimethylbenzene
Molecular Formula
C8H9I
InChI Key
BUNKQJAMHYKQIM-UHFFFAOYSA-N
SMILES
CC1=CC=C(I)C(C)=C1
Appearance (Color)
Clear colorless to pale yellow to yellow to orange
Form
Liquid
Assay (GC)
≥97.5%
Refractive Index
1.5955-1.5995 @ 20?C

Description

4-Iodo-m-xylene is a reactant used in the preparation of paralog-selective Hsp90 inhibitors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Iodo-m-xylene is a reactant used in the preparation of paralog-selective Hsp90 inhibitors.

Solubility
Not miscible or difficult to mix in water.

Notes
Light Sensitive. Protect from light. Store away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text