Thermo Scientific Chemicals

3,5-Di-tert-butyl-4-hydroxybenzoic acid, 98%, Thermo Scientific Chemicals

Catalog number: A16880.18
50 g, Each
Thermo Scientific Chemicals

3,5-Di-tert-butyl-4-hydroxybenzoic acid, 98%, Thermo Scientific Chemicals

Catalog number: A16880.18
50 g, Each
Quantity
Catalog number: A16880.18
also known as A16880-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1421-49-4
Appearance (Color)
Pale yellow to yellow
Assay (Silylated GC)
>97.5%
Assay (Aqueous acid-base Titration)
>97.5 to <102.5%
Form
Powder

Description

3,5-Di-tert-butyl-4-hydroxybenzoic acid was used in the synthesis of antistress agents. It is used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,5-Di-tert-butyl-4-hydroxybenzoic acid was used in the synthesis of antistress agents. It is used as pharmaceutical intermediates.

Solubility
Insoluble in water.

Notes
Store in a cool, dry, conditions in well sealed container. Store away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text