2'-Fluoro-4'-methoxyacetophenone, 99%, Thermo Scientific Chemicals
2'-Fluoro-4'-methoxyacetophenone, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2'-Fluoro-4'-methoxyacetophenone, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number A16891.06
also known as A16891-06
Price (USD)/ Each
37.30
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
37.30
Add to cart
2'-Fluoro-4'-methoxyacetophenone, 99%, Thermo Scientific Chemicals
Catalog numberA16891.06
Price (USD)/ Each
37.30
-
Add to cart
Chemical Identifiers
CAS74457-86-6
IUPAC Name1-(2-fluoro-4-methoxyphenyl)ethan-1-one
Molecular FormulaC9H9FO2
InChI KeyPIRRWUMTIBFCCW-UHFFFAOYSA-N
SMILESCOC1=CC=C(C(C)=O)C(F)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)48.0-54.0?C
FormCrystals or powder or crystalline powder
Appearance (Color)White
Assay (GC)≥98.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only