Thermo Scientific Chemicals

Dimethyl sulfoxide-d{6}, 99.5% (Isotopic), Thermo Scientific Chemicals

Catalog number: A16893.09
10 g, Each
Thermo Scientific Chemicals

Dimethyl sulfoxide-d{6}, 99.5% (Isotopic), Thermo Scientific Chemicals

Catalog number: A16893.09
10 g, Each
Quantity
Catalog number: A16893.09
also known as A16893-09
Price (USD)
Have Questions?

Chemical Identifiers

CAS
2206-27-1
IUPAC Name
(²H₃)methanesulfinyl(²H₃)methane
Molecular Formula
C2H6OS
InChI Key
IAZDPXIOMUYVGZ-WFGJKAKNSA-N
SMILES
[2H]C([2H])([2H])S(=O)C([2H])([2H])[2H]
Appearance (Color)
Clear colorless to pale yellow
Assay (GC)
≥99.5% (UK sourced material)
Form
Liquid
Comment
Material sourced in UK or US
Isotopic purity
≥99.5%

Description

DMSO-d6 is a solvent of choice in NMR spectroscopy, owing to its unique characteristics of dissolving both nonpolar and polar compounds, miscibility with almost all organic solvents and permitting high temperature dynamic NMR studies.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
DMSO-d6 is a solvent of choice in NMR spectroscopy, owing to its unique characteristics of dissolving both nonpolar and polar compounds, miscibility with almost all organic solvents and permitting high temperature dynamic NMR studies.

Solubility
Miscible with water and with almost all organic solvents such as alcohols, esters, ketones, chlorinated solvents and aromatic hydrocarbons.

Notes
NMR solvent. Dimethyl Sulfoxide (DMSO) has a melting point of 20°C, freezing close to room temperature. Sometimes when it is delivered it will be in a solid state. To return the material to a liquid state, thaw it in a warm water bath. Care must be taken to prevent water contamination. It is stable, combustible and moisture sensitive. Incompatible with acid chlorides, strong acids, phosphorus halides, strong oxidizing agents and strong reducing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text