Thermo Scientific Chemicals

4-(Trifluoromethyl)benzyl bromide, 98%, Thermo Scientific Chemicals

Catalog number: A16895.14
25 g, Each
Thermo Scientific Chemicals

4-(Trifluoromethyl)benzyl bromide, 98%, Thermo Scientific Chemicals

Catalog number: A16895.14
25 g, Each
Quantity
Catalog number: A16895.14
also known as A16895-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
402-49-3
IUPAC Name
1-(bromomethyl)-4-(trifluoromethyl)benzene
Molecular Formula
C8H6BrF3
InChI Key
IKSNDOVDVVPSMA-UHFFFAOYSA-N
SMILES
FC(F)(F)C1=CC=C(CBr)C=C1
Form
Crystals or powder or crystalline powder or fused solid
Identification (FTIR)
Conforms
Refractive Index
1.4895-1.4935 @ 20?C
Appearance (Color)
White to cream
Assay (GC)
≥97.5%

Description

4-(Trifluoromethyl)benzyl bromide can be used to produce p-trifluoromethylbenzyl phenyl sulfide. It is used in medicine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(Trifluoromethyl)benzyl bromide can be used to produce p-trifluoromethylbenzyl phenyl sulfide. It is used in medicine.

Solubility
Insoluble in water.

Notes
Store in a cool, dry, conditions in well sealed container. Store away from oxidizing agents. Incompatible with oxidizing agents and bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text