General Description
Lanolin is a waxy material that is composed of long chain esters and lanolin alcohols, acids, and polycarbons.
Application
• As a precursor of histidine, it can be used to enhance the biosynthesis of lovastatin by cultured Aspergillus terreus
• It can be used in stabilizing glass slides during worm-immobilization method
General References: