Thermo Scientific Chemicals

3-Nitrophenyl isocyanate, 97%, Thermo Scientific Chemicals

Catalog number: A16920.06
5 g, Each
Thermo Scientific Chemicals

3-Nitrophenyl isocyanate, 97%, Thermo Scientific Chemicals

Catalog number: A16920.06
5 g, Each
Quantity
Catalog number: A16920.06
also known as A16920-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
3320-87-4
IUPAC Name
1-isocyanato-3-nitrobenzene
Molecular Formula
C7H4N2O3
InChI Key
GFFGYTMCNVMFAJ-UHFFFAOYSA-N
SMILES
[O-][N+](=O)C1=CC=CC(=C1)N=C=O
Melting Point (clear melt)
47.0-57.0?C
Identification (FTIR)
Conforms
Appearance (Color)
Pale yellow to yellow
Assay (GC)
≥96.0%
Form
Fused solid

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text