Thermo Scientific Chemicals

Tetramethylgermanium, 98%, Thermo Scientific Chemicals

Catalog number: A16931.06
5 g, Each
Thermo Scientific Chemicals

Tetramethylgermanium, 98%, Thermo Scientific Chemicals

Catalog number: A16931.06
5 g, Each
Quantity
Catalog number: A16931.06
also known as A16931-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
865-52-1
IUPAC Name
tetramethylgermane
Molecular Formula
C4H12Ge
InChI Key
ZRLCXMPFXYVHGS-UHFFFAOYSA-N
SMILES
C[Ge](C)(C)C
Appearance (Color)
Clear colorless
Assay (GC)
≥97.5%
Form
Liquid
Refractive Index
1.3870-1.3910 @ 20?C

Description

Employed as a Chemical Vapor Deposition (CVD) precursor to germanium oxide thin films.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Employed as a Chemical Vapor Deposition (CVD) precursor to germanium oxide thin films.

Solubility
Not miscible or difficult to mix in water.

Notes
Store away from oxidizing agents. Keep the container tightly sealed and store in cool, dry condition in well ventilated place.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text