Hydrogen chloride, 4M in 1,4-dioxane, 99%, Thermo Scientific Chemicals
Hydrogen chloride, 4M in 1,4-dioxane, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Hydrogen chloride, 4M in 1,4-dioxane, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
0.5 mol
0.1 mol
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number A16935.JT
also known as A16935-JT
Price (USD)/ Each
107.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
126.00 
Save 18.35 (15%)
-
Add to cart
Quantity:
0.5 mol
Request bulk or custom format
Price (USD)/ Each
107.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
126.00 
Save 18.35 (15%)
Add to cart
Hydrogen chloride, 4M in 1,4-dioxane, 99%, Thermo Scientific Chemicals
Catalog numberA16935.JT
Price (USD)/ Each
107.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
126.00 
Save 18.35 (15%)
-
Add to cart
Chemical Identifiers
CAS7647-01-0
IUPAC Namehydrogen chloride
Molecular FormulaClH
InChI KeyVEXZGXHMUGYJMC-UHFFFAOYSA-N
SMILESCl
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Aqueous acid-base Titration)3.6-4.4 mol/L HCl
Appearance (Color)Clear, colorless to very pale yellow
FormLiquid
Water Content (Karl Fischer Titration)≤0.3%
Hydrogen chloride, 4M in 1,4-dioxane is used in the preparation of N-methyl-O-benzoylhydroxylamine hydrochloride via reaction with benzoic acid, carbonyldiimidazole and N-methylhydroxylamine hydrochloride.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only