Thermo Scientific Chemicals

Dibromochloromethane, 97+%, Thermo Scientific Chemicals

Catalog number: A16938.22
100 g, Each
Thermo Scientific Chemicals

Dibromochloromethane, 97+%, Thermo Scientific Chemicals

Catalog number: A16938.22
100 g, Each
Quantity
Catalog number: A16938.22
also known as A16938-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
124-48-1
IUPAC Name
dibromo(chloro)methane
Molecular Formula
CHBr2Cl
InChI Key
GATVIKZLVQHOMN-UHFFFAOYSA-N
SMILES
ClC(Br)Br
Assay (GC)
≥97.0%
Form
Liquid
Refractive Index
1.5455-1.5495 @ 20?C
Appearance (Color)
Clear colorless to yellow

Description

It affords bromochlorocarbene under phase-transfer conditions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It affords bromochlorocarbene under phase-transfer conditions.

Solubility
Not miscible or difficult to mix in water.

Notes
Moisture & Light Sensitive. Store away from light, water/moisture, oxidizing agents and heat. Store under inert gas.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text