5-Aminolevulinic acid hydrochloride, 99%, Thermo Scientific Chemicals
5-Aminolevulinic acid hydrochloride, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

5-Aminolevulinic acid hydrochloride, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mg
2 g
Catalog number A16942.ME
also known as A16942-ME
Price (USD)/ Each
84.65
Online exclusive
94.30 
Save 9.65 (10%)
-
Add to cart
Quantity:
500 mg
Request bulk or custom format
Price (USD)/ Each
84.65
Online exclusive
94.30 
Save 9.65 (10%)
Add to cart
5-Aminolevulinic acid hydrochloride, 99%, Thermo Scientific Chemicals
Catalog numberA16942.ME
Price (USD)/ Each
84.65
Online exclusive
94.30 
Save 9.65 (10%)
-
Add to cart
Chemical Identifiers
CAS5451-09-2
IUPAC Namehydrogen 5-amino-4-oxopentanoic acid chloride
Molecular FormulaC5H10ClNO3
InChI KeyZLHFONARZHCSET-UHFFFAOYSA-N
SMILES[H+].[Cl-].NCC(=O)CCC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
CommentMaterial Sourced in UK and US
Assay (Titration ex Chloride)≥98.5 to ≤101.5% (UK Sourced Material)
Identification (FTIR)Conforms (UK Sourced Material)
Appearance (Color)White to cream
View more
5-Aminolevulinic acid hydrochloride finds an important role as a precursor in the synthesis of tetrapyrroles such as chlorophyll and heme. It is widely utilized in photodynamic therapy of diseases namely, Paget’s disease and human papillomavirus (HPV) infection-associated cervical condylomata acuminata.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in dimethyl sulfoxide, methanol and water.

Notes
Store in a cool place.
RUO – Research Use Only

General References:

  1. Yang, X.; Li, W.; Palasuberniam, P.; Myers, K. A.; Wang, C.; Chen, B. Effects of Silencing Heme Biosynthesis Enzymes on 5-Aminolevulinic Acid-mediated Protoporphyrin IX Fluorescence and Photodynamic Therapy. Photochem. Photobiol. 2015, 91 (4), 923-930.
  2. Nunkaew, T.; Kantachote, D.; Nitoda, T.; Kanzaki, H. Selection of salt tolerant purple nonsulfur bacteria producing 5-aminolevulinic acid (ALA) and reducing methane emissions from microbial rice straw degradation. Appl. Soil Ecol. 2015, 86, 113-120.