It has a wide range of chemical and biochemical roles employing as a regulatory molecule. It acts as a precursor, substrate, or cofactor in diverse biochemical pathways.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It has a wide range of chemical and biochemical roles employing as a regulatory molecule. It acts as a precursor, substrate, or cofactor in diverse biochemical pathways.
Solubility
Soluble in water (4 mg/ml), ethanol (<1mg/mL at 25°C), DMSO (<1mg/mL at 25°C), and HCl (10 mg/ml).
Notes
Store away from oxidizing agents. Keep the container tightly sealed and store in cool, dry condition in well ventilated place.
RUO – Research Use Only
General References:
- Nuki, G. Human purine metabolism: Some recent advances and relationships with immunodeficiency.Ann Rheum Dis.1983,428-11.
- Sonali Jana.; Gyan Singh Shekhawat. Plant growth regulators, adenine sulfate and carbohydrates regulate organogenesis and in vitro flowering of Anethum graveolens.Acta Physiologiae Plantarum,2011,33,(2), 305-311.