Thermo Scientific Chemicals

Adenine sulfate, 98+%

Catalog number: A16964.18
50 g, Each
Thermo Scientific Chemicals

Adenine sulfate, 98+%

Catalog number: A16964.18
50 g, Each
Quantity
Catalog number: A16964.18
also known as A16964-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
321-30-2
IUPAC Name
bis(7H-purin-6-amine); sulfuric acid
Molecular Formula
C10H12N10O4S
InChI Key
LQXHSCOPYJCOMD-UHFFFAOYSA-N
SMILES
OS(O)(=O)=O.NC1=C2NC=NC2=NC=N1.NC1=C2NC=NC2=NC=N1
Form
Powder
Appearance (Color)
White to pale cream
Assay (HPLC)
≥98.0%
Water Content (Karl Fischer Titration)
≤2.0%

Description

It has a wide range of chemical and biochemical roles employing as a regulatory molecule. It acts as a precursor, substrate, or cofactor in diverse biochemical pathways.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It has a wide range of chemical and biochemical roles employing as a regulatory molecule. It acts as a precursor, substrate, or cofactor in diverse biochemical pathways.

Solubility
Soluble in water (4 mg/ml), ethanol (<1mg/mL at 25°C), DMSO (<1mg/mL at 25°C), and HCl (10 mg/ml).

Notes
Store away from oxidizing agents. Keep the container tightly sealed and store in cool, dry condition in well ventilated place.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text