Thermo Scientific Chemicals

Ammonium citrate tribasic, 97+%, Thermo Scientific Chemicals

Catalog number: A16973.22
100 g, Each
Thermo Scientific Chemicals

Ammonium citrate tribasic, 97+%, Thermo Scientific Chemicals

Catalog number: A16973.22
100 g, Each
Quantity
Catalog number: A16973.22
also known as A16973-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
3458-72-8
IUPAC Name
triammonium 2-hydroxypropane-1,2,3-tricarboxylate
Molecular Formula
C6H17N3O7
InChI Key
YWYZEGXAUVWDED-UHFFFAOYSA-N
SMILES
[NH4+].[NH4+].[NH4+].OC(CC([O-])=O)(CC([O-])=O)C([O-])=O
Appearance (Color)
Colorless to white
Impurity content
Iron Fe: 1ppm max.
Impurity content
Lithium Li: 1ppm max.
Impurity content
Sodium Na: 5ppm max.
Particle Size
+70 mesh: ≥90%

Description

Lithium nitrate is used in ceramics, pyrotechnics, salt baths, heat-exchange media, refrigeration systems, rocket propellant and in solar batteries. It acts as an oxidizing agent used in the production of red-colored fireworks and flares, as a catalyst to accelerate the breakdown of nitrogen oxides and as a medium to store heat received from sun for cooking. It is applied to concrete-pavement to withstand weathering effects.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Solubility
Soluble in water. Slightly soluble in alcohol.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text