3-(2-Aminoethylamino)propyltrimethoxysilane, tech. 90%, Thermo Scientific Chemicals
3-(2-Aminoethylamino)propyltrimethoxysilane, tech. 90%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-(2-Aminoethylamino)propyltrimethoxysilane, tech. 90%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
1000 g
Catalog number A16981.18
also known as A16981-18
Price (USD)/ Each
29.90
-
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)/ Each
29.90
Add to cart
3-(2-Aminoethylamino)propyltrimethoxysilane, tech. 90%, Thermo Scientific Chemicals
Catalog numberA16981.18
Price (USD)/ Each
29.90
-
Add to cart
Chemical Identifiers
CAS1760-24-3
IUPAC Namehydrogen (2-aminoethyl)[3-(trimethoxysilyl)propyl]amine chloride
Molecular FormulaC8H23ClN2O3Si
InChI KeySZBIXNNLIKDOEJ-UHFFFAOYSA-N
SMILES[H+].[Cl-].CO[Si](CCCNCCN)(OC)OC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear, colorless
FormLiquid
Assay (GC)>88.0%
Refractive Index1.4400-1.4480 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only