3-(Trifluoromethyl)cinnamic acid, predominantly trans, 98+%, Thermo Scientific Chemicals
3-(Trifluoromethyl)cinnamic acid, predominantly trans, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-(Trifluoromethyl)cinnamic acid, predominantly trans, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number A16983.14
also known as A16983-14
Price (USD)/ Each
156.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
156.00
Add to cart
3-(Trifluoromethyl)cinnamic acid, predominantly trans, 98+%, Thermo Scientific Chemicals
Catalog numberA16983.14
Price (USD)/ Each
156.00
-
Add to cart
Chemical Identifiers
CAS779-89-5
IUPAC Name(2E)-3-[3-(trifluoromethyl)phenyl]prop-2-enoate
Molecular FormulaC10H6F3O2
InChI KeyKSBWHDDGWSYETA-SNAWJCMRSA-M
SMILES[O-]C(=O)C=CC1=CC=CC(=C1)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
FormCrystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)≥98.0 to ≤102.0%
Assay (Silylated GC)≥98.0%
Melting Point (clear melt)131-136?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only