Thermo Scientific Chemicals

(+/-)-2-Octanol, 98%, Thermo Scientific Chemicals

Catalog number: A16985.AP
500 mL, Each
Thermo Scientific Chemicals

(+/-)-2-Octanol, 98%, Thermo Scientific Chemicals

Catalog number: A16985.AP
500 mL, Each
Quantity
Catalog number: A16985.AP
also known as A16985-AP
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
123-96-6
IUPAC Name
octan-2-ol
Molecular Formula
C8H18O
InChI Key
SJWFXCIHNDVPSH-UHFFFAOYNA-N
SMILES
CCCCCCC(C)O
Appearance (Color)
Clear colorless
Form
Liquid
Identification (FTIR)
Conforms
Assay (GC)
≥97.5%
Refractive Index
1.4250-1.4290 @ 20?C

Description

(±)-2-Octanol is used in flavors, paints and coatings, defoaming agent, low-volatility solvent, lubricants, fuels, paints and coatings. It is used as a frother in mineral floatation and wetting agent. It acts as an intermediate in the preparation of surfactants, cosmetic emollients esters and plasticizers. Further, it is also used as a reagent in the synthesis of pharmaceutical compounds and piperine derivatives. In addition to this, it is considered as an alternative of 2-ethylhexanol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(^+)-2-Octanol is used in flavors, paints and coatings, defoaming agent, low-volatility solvent, lubricants, fuels, paints and coatings. It is used as a frother in mineral floatation and wetting agent. It acts as an intermediate in the preparation of surfactants, cosmetic emollients esters and plasticizers. Further, it is also used as a reagent in the synthesis of pharmaceutical compounds and piperine derivatives. In addition to this, it is considered as an alternative of 2-ethylhexanol.

Solubility
Miscible with diethyl ether, acetone, alcohol, most common organic solvents, aromatic and aliphatic hydrocarbons. Slightly miscible with cold water.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text