Thermo Scientific Chemicals

Lithium dihydrogen phosphate, 97%, Thermo Scientific Chemicals

Catalog number: A16987.30
250 g, Each
Thermo Scientific Chemicals

Lithium dihydrogen phosphate, 97%, Thermo Scientific Chemicals

Catalog number: A16987.30
250 g, Each
Quantity
Catalog number: A16987.30
also known as A16987-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
13453-80-0
IUPAC Name
lithium(1+) dihydrogen phosphate
Molecular Formula
H2LiO4P
InChI Key
SNKMVYBWZDHJHE-UHFFFAOYSA-M
SMILES
[Li+].OP(O)([O-])=O
Appearance (Color)
White
Assay (Aqueous acid-base Titration)
≥96.0 to ≤104.0%
Form
Crystals or powder or crystalline powder

Description

Employed as a precursor for LiMX2O7 (M = Fe, V; x =P, As) cathode materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Employed as a precursor for LiMX2O7 (M = Fe, V; x =P, As) cathode materials.

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly sealed and store
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text