Thermo Scientific Chemicals

2-Ethylbutyric acid, 98%, Thermo Scientific Chemicals

Catalog number: A16989.AU
1000 mL, Each
Thermo Scientific Chemicals

2-Ethylbutyric acid, 98%, Thermo Scientific Chemicals

Catalog number: A16989.AU
1000 mL, Each
Quantity
Catalog number: A16989.AU
also known as A16989-AU
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
88-09-5
IUPAC Name
2-ethylbutanoic acid
Molecular Formula
C6H12O2
InChI Key
OXQGTIUCKGYOAA-UHFFFAOYSA-N
SMILES
CCC(CC)C(O)=O
Refractive Index
1.4110-1.4150 @ 20?C
Appearance (Color)
Clear colorless
Assay (Silylated GC)
≥97.5%
Assay (Aqueous acid-base Titration)
≥97.5 to ≤102.5%
Form
Liquid

Description

Diethyl acetic acid (88-09-5) is used in flavoring agent and as an intermediate for pharmaceuticals and dyestuffs. It is also employed in the manufacture of esters and other chemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethyl acetic acid (88-09-5) is used in flavoring agent and as an intermediate for pharmaceuticals and dyestuffs. It is also employed in the manufacture of esters and other chemicals.

Solubility
Sparingly soluble with the usual organic solvents. Solubility in water 18 g/L (20°C).

Notes
Store away from oxidizing agents. Keep the container tightly sealed and store in cool, dry condition in well ventilated place.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text