Thermo Scientific Chemicals

Dibromoacetonitrile, 94%, Thermo Scientific Chemicals

Catalog number: A16994.22
100 g, Each
Thermo Scientific Chemicals

Dibromoacetonitrile, 94%, Thermo Scientific Chemicals

Catalog number: A16994.22
100 g, Each
Quantity
Catalog number: A16994.22
also known as A16994-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
3252-43-5
IUPAC Name
2,2-dibromoacetonitrile
Molecular Formula
C2HBr2N
InChI Key
NDSBDLSWTGLNQA-UHFFFAOYSA-N
SMILES
BrC(Br)C#N
Appearance (Color)
Colorless to yellow to orange
Form
Powder or clear liquid as melt
Refractive Index
1.5385-1.5455 @ 20?C
Assay (GC)
≥92.0%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text