1-Phenylpiperazine hydrochloride, 99%, Thermo Scientific Chemicals
1-Phenylpiperazine hydrochloride, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1-Phenylpiperazine hydrochloride, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number A16995.14
also known as A16995-14
Price (USD)/ Each
58.60
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
58.60
Add to cart
1-Phenylpiperazine hydrochloride, 99%, Thermo Scientific Chemicals
Catalog numberA16995.14
Price (USD)/ Each
58.60
-
Add to cart
Chemical Identifiers
CAS2210-93-7
IUPAC Namehydrogen 1-phenylpiperazine chloride
Molecular FormulaC10H15ClN2
InChI KeyNQNZNONJZASOKL-UHFFFAOYSA-N
SMILES[H+].[Cl-].C1CN(CCN1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream or pink or grey
FormCrystals or powder or crystalline powder
Assay (Titration ex Chloride)≥98.5 to ≤101.5%
Assay (Freebase GC)≥98.5%
Identification (FTIR)Conforms
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only