Thermo Scientific Chemicals

1H-1,2,3-Triazole, 98%, Thermo Scientific Chemicals

Catalog number: A16999.06
5 g, Each
Thermo Scientific Chemicals

1H-1,2,3-Triazole, 98%, Thermo Scientific Chemicals

Catalog number: A16999.06
5 g, Each
Quantity
Catalog number: A16999.06
also known as A16999-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
288-36-8
IUPAC Name
2H-1,2,3-triazole
Molecular Formula
C2H3N3
InChI Key
QWENRTYMTSOGBR-UHFFFAOYSA-N
SMILES
N1N=CC=N1
Assay (GC)
≥97.5%
Form
Liquid
Refractive Index
1.4960-1.5000 @ 20?C
Appearance (Color)
Clear colorless to pale yellow
Identification (FTIR)
Conforms

Description

It effectively promotes the proton conduction in polymer electrolyte membranes via an intermolecular proton-transfer mechanism. A useful triazole for proteomics research

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It effectively promotes the proton conduction in polymer electrolyte membranes via an intermolecular proton-transfer mechanism. A useful triazole for proteomics research

Solubility
Fully miscible in water.

Notes
Store away from oxidizing agents. Keep the container tightly sealed and store in cool, dry condition in well ventilated place.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text