Thermo Scientific Chemicals

5,5-Diphenylhydantoin, 99%, Thermo Scientific Chemicals

Catalog number: A17013.14
25 g, Each
Thermo Scientific Chemicals

5,5-Diphenylhydantoin, 99%, Thermo Scientific Chemicals

Catalog number: A17013.14
25 g, Each
Quantity
Catalog number: A17013.14
also known as A17013-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
57-41-0
IUPAC Name
5,5-diphenylimidazolidine-2,4-dione
Molecular Formula
C15H12N2O2
InChI Key
CXOFVDLJLONNDW-UHFFFAOYSA-N
SMILES
O=C1NC(=O)C(N1)(C1=CC=CC=C1)C1=CC=CC=C1
Form
Powder
Identification (FTIR)
Conforms
Appearance (Color)
White to pale cream
Assay (HPLC)
≥98.5%
Loss on Drying
≤1.0 % (1 g, 105°C)

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text