5,5-Diphenylhydantoin, 99%, Thermo Scientific Chemicals
5,5-Diphenylhydantoin, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

5,5-Diphenylhydantoin, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
Catalog number A17013.36
also known as A17013-36
Price (USD)/ Each
165.00
-
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)/ Each
165.00
Add to cart
5,5-Diphenylhydantoin, 99%, Thermo Scientific Chemicals
Catalog numberA17013.36
Price (USD)/ Each
165.00
-
Add to cart
Chemical Identifiers
CAS57-41-0
IUPAC Name5,5-diphenylimidazolidine-2,4-dione
Molecular FormulaC15H12N2O2
InChI KeyCXOFVDLJLONNDW-UHFFFAOYSA-N
SMILESO=C1NC(=O)C(N1)(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Identification (FTIR)Conforms
Appearance (Color)White to pale cream
Assay (HPLC)≥98.5%
Loss on Drying≤1.0 % (1 g, 105°C)
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only