Thermo Scientific Chemicals

4-tert-Butylphenoxyacetic acid, 98%, Thermo Scientific Chemicals

Catalog number: A17017.06
5 g, Each
Thermo Scientific Chemicals

4-tert-Butylphenoxyacetic acid, 98%, Thermo Scientific Chemicals

Catalog number: A17017.06
5 g, Each
Quantity
Catalog number: A17017.06
also known as A17017-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1798-04-5
IUPAC Name
2-(4-tert-butylphenoxy)acetic acid
Molecular Formula
C12H16O3
InChI Key
FBIGAJNVRFKBJL-UHFFFAOYSA-N
SMILES
CC(C)(C)C1=CC=C(OCC(O)=O)C=C1
Appearance (Color)
White
Melting Point (clear melt)
92.0-98.0?C
Form
Crystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)
≥97.5 to ≤102.5%

Description

Applied as a reagent for the TAC protective group in nucleosides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Applied as a reagent for the TAC protective group in nucleosides.

Solubility
Sparingly Soluble (0.56 g/L) (25°C), Calc.

Notes
Store away from oxidizing agents. Keep the container tightly sealed and store in cool, dry condition in well ventilated place.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text