Applied as a reagent for the TAC protective group in nucleosides.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Applied as a reagent for the TAC protective group in nucleosides.
Solubility
Sparingly Soluble (0.56 g/L) (25°C), Calc.
Notes
Store away from oxidizing agents. Keep the container tightly sealed and store in cool, dry condition in well ventilated place.
RUO – Research Use Only
N D Sinha.; P Davis.; N Usman.; J Pérez.; R Hodge.; J Kremsky.; R Casale. Labile exocyclic amine protection of nucleosides in DNA, RNA and oligonucleotide analog synthesis facilitating N-deacylation, minimizing depurination and chain degradation.Biochimie.,1993,75(1-2), 13-23.
S.N. Podyachev.; I.A. Litvinov.; R.R. Shagidullin.; B.I. Buzykin.; I. Bauer.; D.V. Osyanina.; L.V. Avvakumova.; S.N. Sudakova.; W.D. Habicher.; A.I. Konovalova. Structure and spectroscopic characteristics of 4-tert-butylphenoxyacetylhydrazones of arylaldehydes.Spectrochimica Acta Part A: Molecular and Biomolecular Spectroscopy,2007,66,(2), 250-261.