Thermo Scientific Chemicals

Quinine hemisulfate monohydrate, 98+%, Thermo Scientific Chemicals

Catalog number: A17036.30
250 g, Each
Thermo Scientific Chemicals

Quinine hemisulfate monohydrate, 98+%, Thermo Scientific Chemicals

Catalog number: A17036.30
250 g, Each
Quantity
Catalog number: A17036.30
also known as A17036-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
6119-70-6
IUPAC Name
bis((R)-[(2S,5R)-5-ethenyl-1-azabicyclo[2.2.2]octan-2-yl](6-methoxyquinolin-4-yl)methanol) sulfuric acid dihydrate
Molecular Formula
C40H54N4O10S
InChI Key
ZHNFLHYOFXQIOW-OIGVVMIYNA-N
SMILES
O.O.OS(O)(=O)=O.[H][C@@]1(CN2CCC1C[C@@]2([H])[C@H](O)C1=C2C=C(OC)C=CC2=NC=C1)C=C.[H][C@@]1(CN2CCC1C[C@@]2([H])[C@H](O)C1=C2C=C(OC)C=CC2=NC=C1)C=C
Assay (Non-aqueous acid-base Titration)
≥98.0 to ≤102.0% (Dry wt. basis)
Appearance (Color)
White
Water Content (Karl Fischer Titration)
4.1 to 5.1% (theory 4.6%)
Form
Powder
Optical Rotation
-241° ± 5° (c=2 in 0.1M HCl dry weight basis)

Description

Quinine hemisulfate monohydrate plays a major role in potassium channel blockers. It is also used as an antimalarial, anticholinergic, antihypertensive and a hypoglycemic agent. It inhibits mitochondrial ATP-regulated potassium channel. It is also used to study the metabolism of biocrystalized heme, hemozoin, in malarial parasites and to study the toxicity of heme (FP)-complexes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Quinine hemisulfate monohydrate plays a major role in potassium channel blockers. It is also used as an antimalarial, anticholinergic, antihypertensive and a hypoglycemic agent. It inhibits mitochondrial ATP-regulated potassium channel. It is also used to study the metabolism of biocrystalized heme, hemozoin, in malarial parasites and to study the toxicity of heme (FP)-complexes.

Solubility
Soluble in a mixture of chloroform and absolute alcohol (2:1).

Notes
Light sensitive. Incompatible with strong oxidizing agents, alkalis, ammonia, strong bases and iodine.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text