2-Methylimidazole, 97%, Thermo Scientific Chemicals
2-Methylimidazole, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Methylimidazole, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2500 g
100 g
500 g
Catalog number A17063.0E
also known as A17063-0E
Price (USD)/ Each
321.65
Online exclusive
357.00 
Save 35.35 (10%)
-
Add to cart
Quantity:
2500 g
Request bulk or custom format
Price (USD)/ Each
321.65
Online exclusive
357.00 
Save 35.35 (10%)
Add to cart
2-Methylimidazole, 97%, Thermo Scientific Chemicals
Catalog numberA17063.0E
Price (USD)/ Each
321.65
Online exclusive
357.00 
Save 35.35 (10%)
-
Add to cart
Chemical Identifiers
CAS693-98-1
IUPAC Name2-methyl-1H-imidazole
Molecular FormulaC4H6N2
InChI KeyLXBGSDVWAMZHDD-UHFFFAOYSA-N
SMILESCC1=NC=CN1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)96.0%
Appearance (Color)White to pale yellow or cream
Formcrystalline solid
2-Methylimidazole is used as a raw material for the preparation of nitroimidazole antibiotics, which is useful in combat anaerobic bacterial and parasitic infections. It acts as a ligand in coordination chemistry. It is also employed as a hardener or accelerator for epoxy resin and auxiliary agent for textile dyes. Further, it acts as a catalyst for refolding of enhanced colored fluorescent protein. It is also useful as a building block in the synthesis of biologically active compounds. In addition to this, it is used as an active pharmaceutical ingredient intermediate such as metronidazole and dimetridazole.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Methylimidazole is used as a raw material for the preparation of nitroimidazole antibiotics, which is useful in combat anaerobic bacterial and parasitic infections. It acts as a ligand in coordination chemistry. It is also employed as a hardener or accelerator for epoxy resin and auxiliary agent for textile dyes. Further, it acts as a catalyst for refolding of enhanced colored fluorescent protein. It is also useful as a building block in the synthesis of biologically active compounds. In addition to this, it is used as an active pharmaceutical ingredient intermediate such as metronidazole and dimetridazole.

Solubility
Soluble in water and ethanol.

Notes
Incompatible with strong oxidizing agents, acids, acid chlorides and acid anhydrides.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

General References:

  1. Xu, X. B.; Liu, D. B.; Yu, S. J.; Yu, P.; Zhao, Z. G. Separation and determination of 4-methylimidazole, 2-methylimidazole and 5-hydroxymethylfurfural in beverages by amino trap column coupled with pulsed amperometric detection. Food Chem. 2015, 169, 224-229.
  2. Wu, X.; Huang, M.; Kong, F.; Yu, S. Short communication: Study on the formation of 2-methylimidazole and 4-methylimidazole in the Maillard reaction. J. Dairy Sci. 2015, 98 (12), 8565-8571.