Thermo Scientific Chemicals

2-Methylimidazole, 97%, Thermo Scientific Chemicals

Catalog number: A17063.36
500 g, Each
Thermo Scientific Chemicals

2-Methylimidazole, 97%, Thermo Scientific Chemicals

Catalog number: A17063.36
500 g, Each
Quantity
Catalog number: A17063.36
also known as A17063-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
693-98-1
IUPAC Name
2-methyl-1H-imidazole
Molecular Formula
C4H6N2
InChI Key
LXBGSDVWAMZHDD-UHFFFAOYSA-N
SMILES
CC1=NC=CN1
Assay (GC)
96.0%
Appearance (Color)
White to pale yellow or cream
Form
crystalline solid

Description

2-Methylimidazole is used as a raw material for the preparation of nitroimidazole antibiotics, which is useful in combat anaerobic bacterial and parasitic infections. It acts as a ligand in coordination chemistry. It is also employed as a hardener or accelerator for epoxy resin and auxiliary agent for textile dyes. Further, it acts as a catalyst for refolding of enhanced colored fluorescent protein. It is also useful as a building block in the synthesis of biologically active compounds. In addition to this, it is used as an active pharmaceutical ingredient intermediate such as metronidazole and dimetridazole.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Methylimidazole is used as a raw material for the preparation of nitroimidazole antibiotics, which is useful in combat anaerobic bacterial and parasitic infections. It acts as a ligand in coordination chemistry. It is also employed as a hardener or accelerator for epoxy resin and auxiliary agent for textile dyes. Further, it acts as a catalyst for refolding of enhanced colored fluorescent protein. It is also useful as a building block in the synthesis of biologically active compounds. In addition to this, it is used as an active pharmaceutical ingredient intermediate such as metronidazole and dimetridazole.

Solubility
Soluble in water and ethanol.

Notes
Incompatible with strong oxidizing agents, acids, acid chlorides and acid anhydrides.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text