Thermo Scientific Chemicals

Sodium cholate hydrate, 99%, Thermo Scientific Chemicals

Catalog number: A17074.09
10 g, Each
Thermo Scientific Chemicals

Sodium cholate hydrate, 99%, Thermo Scientific Chemicals

Catalog number: A17074.09
10 g, Each
Quantity
Catalog number: A17074.09
also known as A17074-09
Price (USD)
Have Questions?

Chemical Identifiers

CAS
206986-87-0
IUPAC Name
sodium (4R)-4-[(1R,3aS,3bR,4R,5aS,7R,9aS,9bS,11S,11aR)-4,7,11-trihydroxy-9a,11a-dimethyl-hexadecahydro-1H-cyclopenta[a]phenanthren-1-yl]pentanoate
Molecular Formula
C24H39NaO5
InChI Key
NRHMKIHPTBHXPF-TUJRSCDTSA-M
SMILES
[Na+].[H][C@@]12CC[C@H]([C@H](C)CCC([O-])=O)[C@@]1(C)[C@@H](O)C[C@@]1([H])[C@@]2([H])[C@H](O)C[C@]2([H])C[C@H](O)CC[C@]12C
Loss on Drying
≤6.0% (U.S. specification)
Optical Rotation
33.0 ± 1.5° (c=0.5 in water, non-U.S. specification)
Appearance (Color)
White
Form
Crystals or powder or crystalline powder
Assay (Non-aqueous acid-base Titration)
≥98.5 to ≤101.5% (dry wt. basis, non-U.S. specification)

Description

Biochemical solubilizing agent Sodium cholate hydrate is a non-denaturing detergent used for the extraction of membrane proteins. It is involved in the study to assess infiltration of pressurized promoter solutions in a mesoporous silica. Further, it serves as a biochemical solubilizing agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Biochemical solubilizing agent Sodium cholate hydrate is a non-denaturing detergent used for the extraction of membrane proteins. It is involved in the study to assess infiltration of pressurized promoter solutions in a mesoporous silica. Further, it serves as a biochemical solubilizing agent.

Solubility
Soluble in water.

Notes
Hygroscopic. Incompatible with strong acids.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text