Thermo Scientific Chemicals

3-Indoxyl sulfate potassium salt, 97%, Thermo Scientific Chemicals

Catalog number: A17079.03
1 g, Each
Thermo Scientific Chemicals

3-Indoxyl sulfate potassium salt, 97%, Thermo Scientific Chemicals

Catalog number: A17079.03
1 g, Each
Quantity
Catalog number: A17079.03
also known as A17079-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2642-37-7
IUPAC Name
potassium 1H-indol-3-yl sulfate
Molecular Formula
C8H6KNO4S
InChI Key
MDAWATNFDJIBBD-UHFFFAOYSA-M
SMILES
[K+].[O-]S(=O)(=O)OC1=CNC2=CC=CC=C12
Appearance (Color)
White to pale cream
Form
Crystalline powder
Assay (HPLC)
≥96% (from supplier's CofA)
Assay from Supplier's CofA
≥96.0%
Water Content (Karl Fischer Titration)
≤2.0% (from supplier's CofA)

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text