Thermo Scientific Chemicals

Diethyl 1-propylphosphonate, 97%, Thermo Scientific Chemicals

Catalog number: A17105.09
10 g, Each
Thermo Scientific Chemicals

Diethyl 1-propylphosphonate, 97%, Thermo Scientific Chemicals

Catalog number: A17105.09
10 g, Each
Quantity
Catalog number: A17105.09
Price (USD)
Have Questions?

Chemical Identifiers

CAS
18812-51-6
IUPAC Name
diethyl propylphosphonate
Molecular Formula
C7H17O3P
InChI Key
RUIKOPXSCCGLOM-UHFFFAOYSA-N
SMILES
CCCP(=O)(OCC)OCC
Appearance (Color)
Clear colorless
Form
Liquid
Assay (GC)
≥96.0%
Refractive Index
1.4170-1.4220 @ 20?C

Description

Diethyl 1-propylphosphonate, is used as a medical intermediate and in organic synthesis. It is also used in electronics industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethyl 1-propylphosphonate, is used as a medical intermediate and in organic synthesis. It is also used in electronics industry.

Solubility
Soluble in water (27 g/L) (25°C).

Notes
Stable at room temperature in closed containers under normal storage and handling conditions. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible to oxidizing agents, reducing agents, acids, bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text