3-Benzoyl-1,1,1-trifluoroacetone, is used as a primary and secondary intermediate. It is used as an Intermediate in organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3-Benzoyl-1,1,1-trifluoroacetone, is used as a primary and secondary intermediate. It is used as an Intermediate in organic synthesis.
Solubility
Sparingly Soluble in water (0.24 g/L) (25°C).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from oxidizing agents.
RUO – Research Use Only
H Yokoi. The new triplet-state dimers formed by mixing bis(n-alkylsalicylaldiminato)copper(ii) with certain bis (β-diketonato) copper(ii) complexes in toluene. Chemistry letters. 19702 (9), 1023-1026.
VO Iaroshenko.; D Ostrovskyi.; K Ayub. Synthesis of 4-Trifluoromethylpyridines by [5+ 1] Cyclization of 3-Hydroxy-pent-4-yn-1-ones with Urea. Advanced Synthesis and catalysis. 2013355 (2-3), 576-588.
Chelating ligand.