Thermo Scientific Chemicals

N,N'-Diethylethylenediamine, 96%, Thermo Scientific Chemicals

Catalog number: A17113.06
5 g, Each
Thermo Scientific Chemicals

N,N'-Diethylethylenediamine, 96%, Thermo Scientific Chemicals

Catalog number: A17113.06
5 g, Each
Quantity
Catalog number: A17113.06
also known as A17113-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
111-74-0
Appearance (Color)
Clear, colourless
Form
liquid/low melting solid
Assay (GC)
>95.0%
Water Content (Karl Fischer Titration)
<2.0%
Refractive Index
1.4300-1.4360 @20?C

Description

N,N'-Diethylethylenediamine is used as an internal standard in liquid chromatographic method used for checking the assay of ethambutol. It is utilized for the solvation of lithium hexamethyldisilazide. It acts as a ligand and form coordination complexes such as trans-diaquabis(N,N'-diethylethylenediamine)nickel(II) chloride and copper(II)-N,N-Diethylethylenediamine, which shows thermochromic properties. Further, it is used as a pharmaceutical intermediate used in the synthesis of procainamide hydrochloride, tiapride and cinchocaine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N′-Diethylethylenediamine is used as an internal standard in liquid chromatographic method used for checking the assay of ethambutol. It is utilized for the solvation of lithium hexamethyldisilazide. It acts as a ligand and form coordination complexes such as trans-diaquabis(N,N′-diethylethylenediamine)nickel(II) chloride and copper(II)-N,N-Diethylethylenediamine, which shows thermochromic properties. Further, it is used as a pharmaceutical intermediate used in the synthesis of procainamide hydrochloride, tiapride and cinchocaine.

Solubility
Miscible with water.

Notes
Incompatible with acids, acid chlorides, acid anhydrides, carbon dioxide and strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text