Thermo Scientific Chemicals

N,N'-Diisopropylethylenediamine, 97%, Thermo Scientific Chemicals

Catalog number: A17114.22
100 g, Each
Thermo Scientific Chemicals

N,N'-Diisopropylethylenediamine, 97%, Thermo Scientific Chemicals

Catalog number: A17114.22
100 g, Each
Quantity
Catalog number: A17114.22
also known as A17114-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
4013-94-9
Assay (GC)
≥96.0%
Refractive Index
1.4255-1.4305 @ 20?C
Appearance (Color)
Clear colorless
Form
Liquid

Description

N,N'-Diethylethylenediamine is used in the preparation of 2-imidazolidinecarboxaldehydes and 1,4,6,9-tetraalkyl-1,4,6,9-tetraaza-5,10-dioxaperhydroanthracenes by reacting with glyoxal. It acts as a ligand and form coordination complexes with copper(II), which shows thermochromic properties.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N′-Diethylethylenediamine is used in the preparation of 2-imidazolidinecarboxaldehydes and 1,4,6,9-tetraalkyl-1,4,6,9-tetraaza-5,10-dioxaperhydroanthracenes by reacting with glyoxal. It acts as a ligand and form coordination complexes with copper(II), which shows thermochromic properties.

Solubility
Miscible with water.

Notes
Hygroscopic. Incompatible with strong oxidizing agents and strong acids.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text