Thermo Scientific Chemicals

Phenolphthalein, 98%

Catalog number: A17135.0B
1000 g, Each
Thermo Scientific Chemicals

Phenolphthalein, 98%

Catalog number: A17135.0B
1000 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: A17135.0B
also known as A17135-0B
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
77-09-8
IUPAC Name
3,3-bis(4-hydroxyphenyl)-1,3-dihydro-2-benzofuran-1-one
Molecular Formula
C20H14O4
InChI Key
KJFMBFZCATUALV-UHFFFAOYSA-N
SMILES
OC1=CC=C(C=C1)C1(OC(=O)C2=CC=CC=C12)C1=CC=C(O)C=C1
Appearance (Color)
White to cream
Assay (HPLC)
≥97.5%
Color Change
Colorless to pink @ pH 8.2
Identification (FTIR)
Conforms
lambda-max (UV-Vis)
550-555nm (0.1M NaOH)

Description

Phenolphthalein is widely used as an indicator in acid-base titrations, water treatment applications. Due to its pH sensitivity, It is used to the check extent of carbonation in concrete, soap and in toys as diapering work. It is used in Kastle-Meyer test, which identifies the substances containing blood.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phenolphthalein is widely used as an indicator in acid-base titrations, water treatment applications. Due to its pH sensitivity, It is used to the check extent of carbonation in concrete, soap and in toys as diapering work. It is used in Kastle-Meyer test, which identifies the substances containing blood.

Solubility
Soluble in alcohol. Slightly soluble in dimethyl sulfoxide and insoluble in benzene or hexane.

Notes
Acid-base indicator: pH 8.0 - 10.0.Incompatible with strong oxidizing agents.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text