Thermo Scientific Chemicals

2-Adamantanamine hydrochloride, 98+%, Thermo Scientific Chemicals

Catalog number: A17140.14
25 g, Each
Thermo Scientific Chemicals

2-Adamantanamine hydrochloride, 98+%, Thermo Scientific Chemicals

Catalog number: A17140.14
25 g, Each
Quantity
Catalog number: A17140.14
also known as A17140-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10523-68-9
IUPAC Name
hydrogen adamantan-2-amine chloride
Molecular Formula
C10H18ClN
InChI Key
WLDWDRZITJEWRJ-UHFFFAOYSA-N
SMILES
[H+].[Cl-].NC1C2CC3CC(C2)CC1C3
Assay (Titration ex Chloride)
≥98.0 to ≤102.0%
Appearance (Color)
White
Form
Crystals or powder or crystalline powder

Description

2-Adamantylamine hydrochloride was used to prepare 2-adamantylamide of 2?-(carboxymethoxime)-olivomycin I. The HPLC assay of 2-adamantylamine hydrochloride after pre-column derivatization with 4-fluoro-7-nitro-2,1,3-benzoxadiazole has been studied. It is also used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Adamantylamine hydrochloride was used to prepare 2-adamantylamide of 2′-(carboxymethoxime)-olivomycin I. The HPLC assay of 2-adamantylamine hydrochloride after pre-column derivatization with 4-fluoro-7-nitro-2,1,3-benzoxadiazole has been studied. It is also used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text